missing translation for 'onlineSavingsMsg'
Learn More

cis-3-(2-Chloro-3,3,3-trifluoro-1-propen-1-yl)-2,2-dimethyl-cyclopropanemethanol, TRC

Product Code. 30359414
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359414 100 mg 100mg
30354961 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30359414 Supplier Toronto Research Chemicals Supplier No. TRCC422020100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C9 H12 Cl F3 O
Molecular Weight (g/mol) 228.639
Synonym (1R,3R)-rel-3-(2-Chloro-3,3,3-trifluoro-1-propen-1-yl)-2,2-dimethyl-cyclopropanemethanol
IUPAC Name [(1R,3R)-3-[(Z)-2-chloro-3,3,3-trifluoroprop-1-enyl]-2,2-dimethylcyclopropyl]methanol
SMILES CC1(C)[C@H](CO)[C@@H]1\C=C(/Cl)\C(F)(F)F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H12ClF3O/c1-8(2)5(6(8)4-14)3-7(10)9(11,12)13/h3,5-6,14H,4H2,1-2H3/b7-3-/t5-,6+/m0/s1
Formula Weight 228.053
Chemical Name or Material cis-3-(2-Chloro-3,3,3-trifluoro-1-propen-1-yl)-2,2-dimethyl-cyclopropanemethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.