missing translation for 'onlineSavingsMsg'
Learn More
Learn More
13-cis Retinoic Acid-d5 Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 13-cis-methyl Retinoate-2',2',2',3,3-d5 |
| Molecular Formula | C21 2H5 H25 O2 |
| Quantity | 1 mg |
| Synonym | 13-cis-Methyl retinoate-2',2',2',3,3-D5, 13-cis-Methyl retinoate-d5, Methyl 13-cis-retinoate-(2-methyl-d3,3,3-d2), 13-cis-Retinoic acid methyl ester-d5, 13-cis-Retinoic acid-d5 methyl ester |
| InChI Formula | InChI=1S/C21H30O2/c1-16(9-7-10-17(2)15-20(22)23-6)12-13-19-18(3)11-8-14-21(19,4)5/h7,9-10,12-13,15H,8,11,14H2,1-6H3/b10-7+,13-12+,16-9+,17-15-/i3D3,11D2 |
| SMILES | [2H]C([2H])([2H])C1=C(\C=C\C(=C\C=C\C(=C/C(=O)OC)\C)\C)C(C)(C)CCC1([2H])[2H] |
| IUPAC Name | methyl (2Z,4E,6E,8E)-9-[3,3-dideuterio-6,6-dimethyl-2-(trideuteriomethyl)cyclohexen-1-yl]-3,7-dimethylnona-2,4,6,8-tetraenoate |
| Molecular Weight (g/mol) | 319.49 |
| Formula Weight | 319.256 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?