missing translation for 'onlineSavingsMsg'
Learn More

beta-Cyclocitral, Technical Grade, TRC

Product Code. 30359686
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359686 1 g -
30354311 10 g -
30362628 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30359686 Supplier Toronto Research Chemicals Supplier No. TRCC9878251G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 432-25-7
Molecular Formula C10 H16 O
Molecular Weight (g/mol) 152.23
Synonym 1-Cyclohexene-1-carboxaldehyde, 2,6,6-trimethyl-, 2,6,6-Trimethyl-1-cyclohexene-1-carboxaldehyde, 1-Formyl-2,6,6-trimethyl-1-cyclohexene, 2,6,6-Trimethyl-1-cyclohexenecarboxaldehyde, β-Cyclocitral
IUPAC Name 2,6,6-trimethylcyclohexene-1-carbaldehyde
SMILES CC1=C(C=O)C(C)(C)CCC1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H16O/c1-8-5-4-6-10(2,3)9(8)7-11/h7H,4-6H2,1-3H3
Formula Weight 152.1201
Chemical Name or Material beta-Cyclocitral
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.