missing translation for 'onlineSavingsMsg'
Learn More

Cresyl Diphenyl Phosphate-d7 Isomer 1, TRC

Product Code. 30359929
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359929 25 mg -
30355852 5 mg -
30357193 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30359929 Supplier Toronto Research Chemicals Supplier No. TRCC78301625MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C19 D7 H10 O4 P
Molecular Weight (g/mol) 347.353
Synonym Cresyl DPP-d7, Cresyl Diphenyl Phosphate-d7, Diphenyl Cresol Phosphate-d7, Diphenyl Cresyl Phosphate-d7, Diphenyl Tolyl Phosphate-d7, Disflamoll DPK-d7, Durad CDP-d7, Kronitex CDP-d7, Methylphenyl Diphenyl Phosphate-d7, Monocresyl Diphenyl Phosphate-d7, Phosflex 112-d7, Phosflex CDP-d7, Phosphoric Acid Cresyl Diphenyl Ester-d7, Tolyl Diphenyl Phosphate-d7
IUPAC Name diphenyl [2,3,4,6-tetradeuterio-5-(trideuteriomethyl)phenyl] phosphate
SMILES [2H]c1c([2H])c(OP(=O)(Oc2ccccc2)Oc3ccccc3)c([2H])c(c1[2H])C([2H])([2H])[2H]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H17O4P/c1-16-9-8-14-19(15-16)23-24(20,21-17-10-4-2-5-11-17)22-18-12-6-3-7-13-18/h2-15H,1H3/i1D3,8D,9D,14D,15D
Formula Weight 347.13
Chemical Name or Material Cresyl Diphenyl Phosphate-d7 Isomer 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.