missing translation for 'onlineSavingsMsg'
Learn More

S-Desmethyl S-Chloromethyl Dronedarone, TRC

Product Code. 30359974
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359974 100 mg 100mg
30362338 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30359974 Supplier Toronto Research Chemicals Supplier No. TRCD291470100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1520080-11-8
Molecular Formula C31H43ClN2O5S
Molecular Weight (g/mol) 591.2
Synonym N-[2-Butyl-3-[4-[3-(dibutylamino)propoxy]benzoyl]-5-benzofuranyl]-1-chloro-methanesulfonamide
SMILES CCCCN(CCCC)CCCOc1ccc(cc1)C(=O)c2c(CCCC)oc3ccc(NS(=O)(=O)CCl)cc23

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C31H43ClN2O5S/c1-4-7-11-29-30(27-22-25(14-17-28(27)39-29)33-40(36,37)23-32)31(35)24-12-15-26(16-13-24)38-21-10-20-34(18-8-5-2)19-9-6-3/h12-17,22,33H,4-11,18-21,23H2,1-3H3
Formula Weight 590.2581
Chemical Name or Material S-Desmethyl S-Chloromethyl Dronedarone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.