missing translation for 'onlineSavingsMsg'
Learn More

Cefixime Methyl tert-Butyl Diester, TRC

Product Code. 30360325
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360325 10 mg 10mg
30360748 5 mg 5mg
30367462 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30360325 Supplier Toronto Research Chemicals Supplier No. TRCC24281310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C21 H25 N5 O7 S2
Molecular Weight (g/mol) 523.582
IUPAC Name tert-butyl (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(2-methoxy-2-oxoethoxy)iminoacetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES COC(=O)CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)OC(C)(C)C)C=C)\c3csc(N)n3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H25N5O7S2/c1-6-10-8-34-18-14(17(29)26(18)15(10)19(30)33-21(2,3)4)24-16(28)13(11-9-35-20(22)23-11)25-32-7-12(27)31-5/h6,9,14,18H,1,7-8H2,2-5H3,(H2,22,23)(H,24,28)/b25-13-/t14-,18-/m1/s1
Formula Weight 523.12
Chemical Name or Material Cefixime Methyl tert-Butyl Diester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.