missing translation for 'onlineSavingsMsg'
Learn More

N-[[(3E)-2-Chloro-3-[(phenylamino)methylene]-1-cyclopenten-1-yl]methylene]-benzenamine Hydrochloride, TRC

Product Code. 30362167
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362167 250 mg 250mg
30351726 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30362167 Supplier Toronto Research Chemicals Supplier No. TRCC990680250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1448171-86-5
Molecular Formula C19H17ClN2 . HCl
Molecular Weight (g/mol) 345.3
Synonym Benzenamine, N-[[(3E)-2-chloro-3-[(phenylamino)methylene]-1-cyclopenten-1-yl]methylene]-, hydrochloride (1:1), [N(E)]- (ACI), [N(E)]-N-[[(3E)-2-Chloro-3-[(phenylamino)methylene]-1-cyclopenten-1-yl]methylene]benzenamine hydrochloride, N-[[(3E)-2-Chloro-3-[(phenylamino)methylene]-1-cyclopenten-1-yl]methylene]benzenamine hydrochloride, N-[(E)-[2-Chloro-3-[(E)-phenyliminomethyl]cyclopent-2-en-1-ylidene]methyl]aniline hydrochloride
IUPAC Name N-[(E)-[2-chloro-3-(phenyliminomethyl)cyclopent-2-en-1-ylidene]methyl]aniline;hydrochloride
SMILES Cl.ClC1=C(CC/C/1=C\Nc2ccccc2)\C=N\c3ccccc3

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C19H17ClN2.ClH/c20-19-15(13-21-17-7-3-1-4-8-17)11-12-16(19)14-22-18-9-5-2-6-10-18;/h1-10,13-14,21H,11-12H2;1H/b15-13+,22-14?;
Chemical Name or Material N-[((3E)-2-Chloro-3-[(phenylamino)methylene]-1-cyclopenten-1-yl)methylene]-benzenamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.