missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Z)-1-Chloro-2-nonene, TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
Specifications
Specifications
| Chemical Name or Material | (Z)-1-Chloro-2-nonene |
| CAS | 67242-75-5 |
| Molecular Formula | C9H17Cl |
| Quantity | 2.5 mg |
| Synonym | (2Z)-1-Chloro-2-nonene |
| InChI Formula | InChI=1S/C9H17Cl/c1-2-3-4-5-6-7-8-9-10/h7-8H,2-6,9H2,1H3/b8-7- |
| SMILES | CCCCCC\C=C/CCl |
| IUPAC Name | (Z)-1-chloronon-2-ene |
| Molecular Weight (g/mol) | 160.68 |
| Formula Weight | 160.1019 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction