missing translation for 'onlineSavingsMsg'
Learn More

Cetirizine (2,2-Dimethyl-1,3-dioxolan-4-yl)methyl ester, TRC

Product Code. 30364457
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364457 25 mg 25mg
30353289 100 mg 100mg
30354862 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364457 Supplier Toronto Research Chemicals Supplier No. TRCC28114525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C27 H35 Cl N2 O5
Molecular Weight (g/mol) 503.03
IUPAC Name (2,2-dimethyl-1,3-dioxolan-4-yl)methyl 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetate
SMILES CC1(C)OCC(COC(=O)COCCN2CCN(CC2)C(c3ccccc3)c4ccc(Cl)cc4)O1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C27H35ClN2O5/c1-27(2)34-19-24(35-27)18-33-25(31)20-32-17-16-29-12-14-30(15-13-29)26(21-6-4-3-5-7-21)22-8-10-23(28)11-9-22/h3-11,24,26H,12-20H2,1-2H3
Formula Weight 502.223
Chemical Name or Material Cetirizine (2,2-Dimethyl-1,3-dioxolan-4-yl)methyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.