missing translation for 'onlineSavingsMsg'
Learn More

Cinacalcet, TRC

Product Code. 30364497
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364497 5 mg 5mg
30357482 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30364497 Supplier Toronto Research Chemicals Supplier No. TRCC4417905MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 226256-56-0
Molecular Formula C22 H22 F3 N
Molecular Weight (g/mol) 357.41
Synonym 1-Naphthalenemethanamine, α-methyl-N-[3-[3-(trifluoromethyl)phenyl]propyl]-, (αR)-, (αR)-α-Methyl-N-[3-[3-(trifluoromethyl)phenyl]propyl]-1-naphthalenemethanamine, AMG 073, Cinacalcet, N-[(1R)-1-(1-Naphthyl)ethyl]-3-[3-(trifluoromethyl)phenyl]propan-1-amine
IUPAC Name N-[(1R)-1-naphthalen-1-ylethyl]-3-[3-(trifluoromethyl)phenyl]propan-1-amine
SMILES C[C@@H](NCCCc1cccc(c1)C(F)(F)F)c2cccc3ccccc23

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C22H22F3N/c1-16(20-13-5-10-18-9-2-3-12-21(18)20)26-14-6-8-17-7-4-11-19(15-17)22(23,24)25/h2-5,7,9-13,15-16,26H,6,8,14H2,1H3/t16-/m1/s1
Formula Weight 357.1704
Chemical Name or Material Cinacalcet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.