missing translation for 'onlineSavingsMsg'
Learn More

p-Cresol Glucuronide-d7, TRC

Product Code. 30366555
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366555 2.5 mg 2.5mg
30364792 25 mg 25mg
30355283 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30366555 Supplier Toronto Research Chemicals Supplier No. TRCC7820072.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material p-Cresol Glucuronide-d7
Purity Grade Notes HPLC
Molecular Formula C13D7H9O7
Quantity 2.5 mg
Synonym 4-Methylphenyl β-D-Glucopyranosiduronic Acid-d7, β-D-p-Tolyl-glucopyranosiduronic Acid, -d7 p-Tolyl-β-D-glucosiduronic Acid-d7, 4-Cresol Glucuronide-d7, p-Methylphenyl-β-D-glucopyranosiduronic Acid-d7, p-Tolyl-β-glucuronic Acid-d7
InChI Formula InChI=1S/C13H16O7/c1-6-2-4-7(5-3-6)19-13-10(16)8(14)9(15)11(20-13)12(17)18/h2-5,8-11,13-16H,1H3,(H,17,18)/t8-,9-,10?,11?,13+/m0/s1/i1D3,2D,3D,4D,5D
SMILES [2H]c1c([2H])c(c([2H])c([2H])c1O[C@@H]2OC([C@@H](O)[C@H](O)C2O)C(=O)O)C([2H])([2H])[2H]
IUPAC Name (3S,4S,6S)-3,4,5-trihydroxy-6-[2,3,5,6-tetradeuterio-4-(trideuteriomethyl)phenoxy]tetrahydropyran-2-carboxylic acid
Molecular Weight (g/mol) 291.3
Formula Weight 291.1335
Percent Purity >95
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.