missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diamino-4,5-ethylenedioxybenzene, Dihydrochloride, TRC

Product Code. 30367219
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367219 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367219 Supplier Toronto Research Chemicals Supplier No. TRCD41632025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Diamino-4,5-ethylenedioxybenzene, Dihydrochloride
CAS 73448-02-9
Molecular Formula C8 H10 N2 O2
Quantity 25 mg
Synonym 1,4-Benzodioxin-6,7-diamine, 2,3-dihydro-, hydrochloride (1:2), 1,4-Benzodioxin-6,7-diamine, 2,3-dihydro-, dihydrochloride (9CI), 2,3-Dihydro-1,4-benzodioxin-6,7-diamine dihydrochloride, 1,2-Diamino-4,5-ethylenedioxybenzene dihydrochloride, 2,3-Dihydrobenzo[1,4]dioxine-6,7-diamine dihydrochloride
InChI Formula InChI=1S/C8H10N2O2/c9-5-3-7-8(4-6(5)10)12-2-1-11-7/h3-4H,1-2,9-10H2
SMILES Nc1cc2OCCOc2cc1N
IUPAC Name 2,3-dihydro-1,4-benzodioxine-6,7-diamine
Molecular Weight (g/mol) 166.18
Formula Weight 166.0742
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.