missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroindole-3-acetic Acid, TRC

Product Code. 30367230
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367230 250 mg -
30352963 1 g 1g
30359298 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30367230 Supplier Toronto Research Chemicals Supplier No. TRCC367020250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2519-61-1
Molecular Formula C10 H8 Cl N O2
Molecular Weight (g/mol) 209.63
Synonym 4-Chloro-1H-indole-3-acetic acid (ACI), Indole-3-acetic acid, 4-chloro- (7CI, 8CI), 2-(4-Chloro-1H-indol-3-yl)acetic acid, 4-Chloro-IAA, 4-Chloroindole-3-acetic acid, 4-Chloroindoleacetic acid, 4-Chloroindolyl-3-acetic acid, NSC 295294, 4-Chloroindole-3-acetic Acid
IUPAC Name 2-(4-chloro-1H-indol-3-yl)acetic acid
SMILES OC(=O)Cc1c[nH]c2cccc(Cl)c12

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C10H8ClNO2/c11-7-2-1-3-8-10(7)6(5-12-8)4-9(13)14/h1-3,5,12H,4H2,(H,13,14)
Formula Weight 209.0244
Chemical Name or Material 4-Chloroindole-3-acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.