missing translation for 'onlineSavingsMsg'
Learn More

N-Desformyl-4-desacetyl Vincristine Disulfate Salt (>80%), TRC

Product Code. 30368398
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368398 1 mg 1mg
30358375 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30368398 Supplier Toronto Research Chemicals Supplier No. TRCD2899301MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C43 H54 N4 O8 . H2 S O4
Molecular Weight (g/mol) 951.07
Synonym O4-Deacetyl-1-demethyl-vincaleukoblastine Disulfate Salt, Desacetyl Desformyl Vincristine Disulfate Salt
IUPAC Name methyl (3aR,3a1R,4R,5S,5aR,10bS)-3a-ethyl-9-((3S,5S,7S,9S)-5-ethyl-5-hydroxy-9-(methoxycarbonyl)-1,4,5,6,7,8,9,10-octahydro-2H-3,7-methano[1]azacycloundecino[5,4-b]indol-9-yl)-4,5-dihydroxy-8-methoxy-3a,3a1,4,5,5a,6,11,12-octahydro-1H-indolizino[8,1-cd]carbazole-5-carboxylate bis(sulfate)
SMILES CC[C@]12[C@]3([C@]4(C5=CC([C@]6(C(OC)=O)C7=C(C8=CC=CC=C8N7)CC[N@]9C[C@@]([H])(C6)C[C@](C9)(CC)O)=C(C=C5N[C@]4([C@]([C@@H]2O)(C(OC)=O)O)[H])OC)CCN3CC=C1)[H].O=S(O)(O)=O.O=S(O)(O)=O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C43H54N4O8.2H2O4S/c1-6-39(51)21-25-22-42(37(49)54-4,33-27(13-17-46(23-25)24-39)26-11-8-9-12-30(26)44-33)29-19-28-31(20-32(29)53-3)45-34-41(28)15-18-47-16-10-14-40(7-2,35(41)47)36(48)43(34,52)38(50)55-5;2*1-5(2,3)4/h8-12,14,19-20,25,34-36,44-45,48,51-52H,6-7,13,15-18,21-24H2,1-5H3;2*(H2,1,2,3,4)/t25-,34-,35+,36-,39+,40-,41+,42+,43+;;/m1../s1
Formula Weight 950.33
Chemical Name or Material N-Desformyl-4-desacetyl Vincristine Disulfate Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.