missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylhexyl Salicylate-d4, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Ethylhexyl Salicylate-d4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C15H18D4O3 |
| Quantity | 1 mg |
| Synonym | 2-Ethylhexyl o-Hydroxybenzoate-d4, Dermoblock OS-d4, Escalol 587-d4, Eusolex OS-d4, HallBrite OS-USP-d4, NSC 46151-d4, Neo Heliopan OS-d4, Octisalate-d4, Octyl Salicylate-d4, Parsol EHS-d4, 2-Ethylhexyl Ester Salicylic Acid-d4, Solarom OS-d4, Sunarome O-d4, Sunarome WMO-d4, Uvinul 0-18-d4, WMO-d4 |
| InChI Formula | InChI=1S/C15H22O3/c1-3-5-8-12(4-2)11-18-15(17)13-9-6-7-10-14(13)16/h6-7,9-10,12,16H,3-5,8,11H2,1-2H3/i6D,7D,9D,10D |
| SMILES | OC1=C([2H])C([2H])=C([2H])C([2H])=C1C(OCC(CCCC)CC)=O |
| Molecular Weight (g/mol) | 254.36 |
| Percent Purity | >95 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?