missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 3,5-Dimethoxybenzylphosphonate, TRC

Product Code. 30368927
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368927 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30368927 Supplier Toronto Research Chemicals Supplier No. TRCD4442602.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl 3,5-Dimethoxybenzylphosphonate
CAS 108957-75-1
Molecular Formula C13 H21 O5 P
Quantity 2.5 g
InChI Formula InChI=1S/C13H21O5P/c1-5-17-19(14,18-6-2)10-11-7-12(15-3)9-13(8-11)16-4/h7-9H,5-6,10H2,1-4H3
SMILES CCOP(=O)(Cc1cc(OC)cc(OC)c1)OCC
IUPAC Name 1-(diethoxyphosphorylmethyl)-3,5-dimethoxybenzene
Molecular Weight (g/mol) 288.28
Formula Weight 288.1127
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.