missing translation for 'onlineSavingsMsg'
Learn More

Ethyl-2,2,2-d3 Acetate, TRC

Product Code. 30370436
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30370436 10 mg
30371241 50 mg
30383753 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30370436 Supplier Toronto Research Chemicals Supplier No. TRCE67836110MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109545-01-9
Molecular Formula C4 2H3 H5 O2
Molecular Weight (g/mol) 91.12
Synonym Ethan-2,2,2-d3-ol, acetate (9CI), Ethyl-2,2,2-d3 acetate (6CI), Acetic acid ethyl-2,2,2-d3 ester, Ethyl-2,2,2-D3 acetate, Ethyl acetate-d3
IUPAC Name 2,2,2-trideuterioethyl acetate
SMILES [2H]C([2H])([2H])COC(=O)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3/i1D3
Formula Weight 91.0713
Chemical Name or Material Ethyl-2,2,2-D3 Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.