missing translation for 'onlineSavingsMsg'
Learn More

Dihydro a-Ergocryptine, TRC

Product Code. 30370467
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
5 mg
Unit Size:
0.5mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30370467 5 mg -
30388365 2.5 mg 2.5mg
30386617 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30370467 Supplier Toronto Research Chemicals Supplier No. TRCD4485605MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25447-66-9
Molecular Formula C32 H43 N5 O5
Molecular Weight (g/mol) 577.71
Synonym Ergotaman-3',6',18-trione, 9,10-dihydro-12'-hydroxy-2'-(1-methylethyl)-5'-(2-methylpropyl)-, (5'α,10α)-, Ergocryptine, dihydro- (7CI,8CI), (5'α,10α)-9,10-Dihydro-12'-hydroxy-2'-(1-methylethyl)-5'-(2-methylpropyl)ergotaman-3',6',18-trione, 8H-Oxazolo[3,2-a]pyrrolo[2,1-c]pyrazine, ergotaman-3',6',18-trione deriv., Indolo[4,3-fg]quinoline, ergotaman-3',6',18-trione deriv., 9,10-Dihydro-α-ergocryptine, 9,10-Dihydroergocryptine, 9,10-Dihydroergokryptine, Almirid, Dihydro-α-ergocriptine, Dihydro-α-ergocryptin, Dihydro-α-ergocryptine, Dihydroergocriptine, Dihydroergocryptine, α-Dihydroergokryptine
SMILES CC(C)C[C@@H]1N2C(=O)[C@](NC(=O)[C@@H]3C[C@H]4[C@@H](Cc5c[nH]c6cccc4c56)N(C)C3)(O[C@@]2(O)[C@@H]7CCCN7C1=O)C(C)C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C32H43N5O5/c1-17(2)12-25-29(39)36-11-7-10-26(36)32(41)37(25)30(40)31(42-32,18(3)4)34-28(38)20-13-22-21-8-6-9-23-27(21)19(15-33-23)14-24(22)35(5)16-20/h6,8-9,15,17-18,20,22,24-26,33,41H,7,10-14,16H2,1-5H3,(H,34,38)/t20-,22-,24-,25+,26+,31-,32+/m1/s1
Formula Weight 577.3264
Chemical Name or Material Dihydro alpha-Ergocryptine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.