missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disperse Diazo Black 3BF, TRC
Specifications
Specifications
| Chemical Name or Material | Disperse Black 2 |
| CAS | 6232-57-1 |
| Molecular Formula | C14 H16 N4 O |
| Quantity | 2.5 g |
| Synonym | Aniline, 2-methoxy-5-methyl-4,4'-azodi- (7CI), Benzenamine, 4-[(4-aminophenyl)azo]-2-methoxy-5-methyl- (9CI), 4-[2-(4-Aminophenyl)diazenyl]-2-methoxy-5-methylbenzenamine, 1-(4-Aminophenylazo)-2-methyl-4-amino-5-methoxybenzene, 4,4'-Diamino-2-methyl-5-methoxyazobenzene, 4,4'-Diamino-2'-methyl-5'-methoxy-1,1'-azobenzene, Acetamine Diazo Black 3B, Acetoquinone Diazo Navy 2BZ, Amacel Developed Black 3BS, C.I. 11255, C.I. Disperse Black 2, Cibacet Diazo Black B, Diazacyl Black B, Disperse Diazo Black, Disperse Diazo Black 3BF, Dispersol Black 2B, Dispersol Diazo Black 2BN, Fast Black C Base, Kayalon Diazo Black BBS, Meisei Acemyl Diazo Black 3BF, Meisei Teryl Diazo Navy CN, Meisei Teryl Diazo Navy H3B, Naphtoelan Black D Base, Novalon Diazo Black B, Nyloquinone Diazo Navy 2B, Serisol Diazo Black 2B, Setacyl Diazo Black B, Sumikaron Diazo Navy 3B, Terasil Diazo Black B |
| InChI Formula | InChI=1S/C14H16N4O/c1-9-7-12(16)14(19-2)8-13(9)18-17-11-5-3-10(15)4-6-11/h3-8H,15-16H2,1-2H3/b18-17+ |
| SMILES | COc1cc(N=Nc2ccc(N)cc2)c(C)cc1N |
| IUPAC Name | 4-[(E)-(4-aminophenyl)diazenyl]-2-methoxy-5-methylaniline |
| Molecular Weight (g/mol) | 256.3 |
| Formula Weight | 256.1324 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?