missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl Phosphite, TRC

Product Code. 30371905
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30371905 5 g 5g
30378522 10 g -
30378038 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30371905 Supplier Toronto Research Chemicals Supplier No. TRCD4295155G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13086-84-5
Molecular Formula C8H19O3P
Molecular Weight (g/mol) 194.21
Synonym Phosphonic Acid Bis(1,1-dimethylethyl) Ester, Phosphonic Acid Di-tert-butyl Ester, Di-tert-butyl Phosphonate
IUPAC Name 2-methyl-2-[(2-methylpropan-2-yl)oxyphosphonoyloxy]propane
SMILES CC(C)(C)OP(=O)OC(C)(C)C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H19O3P/c1-7(2,3)10-12(9)11-8(4,5)6/h12H,1-6H3
Formula Weight 194.1072
Chemical Name or Material Di-tert-butyl Phosphite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.