missing translation for 'onlineSavingsMsg'
Learn More

Dicumarol, TRC

Product Code. 30372391
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372391 5 g -
30378427 2.5 g 2.5g
30370165 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30372391 Supplier Toronto Research Chemicals Supplier No. TRCD4365905G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-76-2
Molecular Formula C19 H12 O6
Molecular Weight (g/mol) 336.3
Synonym Coumarin, 3,3'-methylenebis[4-hydroxy-, 3,3'-Methylenebis[4-hydroxy-1,2-benzopyrone], 3,3'-Methylenebis[4-hydroxycoumarin], Acadyl, Acavyl, Antitrombosin, Baracoumin, Bis(4-hydroxycoumarin-3-yl)methane, Bis-3,3'-(4-hydroxycoumarinyl)methane, Bishydroxycoumarin, Cuma, Cumid, Di-4-hydroxy-3,3'-methylenedicoumarin, Dicoumal, Dicoumarin, Dicoumarol, Dicuman, Dicumarine, Dicumarol, Dicumol, Dufalone, Kumoran, Melitoxin, NC 034, NSC 17860, NSC 221570, NSC 41834, Temparin, Trombosan
IUPAC Name 4-hydroxy-3-[(4-hydroxy-2-oxochromen-3-yl)methyl]chromen-2-one
SMILES OC1=C(CC2=C(O)c3ccccc3OC2=O)C(=O)Oc4ccccc14

Specifications

Quantity 5 g
InChI Formula InChI=1S/C19H12O6/c20-16-10-5-1-3-7-14(10)24-18(22)12(16)9-13-17(21)11-6-2-4-8-15(11)25-19(13)23/h1-8,20-21H,9H2
Formula Weight 336.0634
Chemical Name or Material Dicoumarol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.