missing translation for 'onlineSavingsMsg'
Learn More

2,2-dichloro-1-(4-chlorophenyl)ethanone, TRC

Product Code. 30372502
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30372502 50 mg
30381659 100 mg
30373416 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30372502 Supplier Toronto Research Chemicals Supplier No. TRCD65050050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5157-57-3
Molecular Formula C8H5OCl3
Molecular Weight (g/mol) 223.48
IUPAC Name 2,2-dichloro-1-(4-chlorophenyl)ethan-1-one
SMILES O=C(C1=CC=C(Cl)C=C1)C(Cl)Cl

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C8H5Cl3O/c9-6-3-1-5(2-4-6)7(12)8(10)11/h1-4,8H
Formula Weight 221.94
Chemical Name or Material 2,2-dichloro-1-(4-chlorophenyl)ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.