missing translation for 'onlineSavingsMsg'
Learn More

N-Ethoxycarbonyl Dabigatran Ethyl Ester, TRC

Product Code. 30373747
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30373747 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373747 Supplier Toronto Research Chemicals Supplier No. TRCE6782705MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1416446-40-6
Molecular Formula C30H33N7O5
Quantity 5 mg
Synonym N-[[2-[[[4-[[(Ethoxycarbonyl)amino]iminomethyl]phenyl]amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-beta-alanine Ethyl Ester
InChI Formula InChI=1S/C30H33N7O5/c1-4-41-27(38)15-17-37(25-8-6-7-16-32-25)29(39)21-11-14-24-23(18-21)34-26(36(24)3)19-33-22-12-9-20(10-13-22)28(31)35-30(40)42-5-2/h6-14,16,18,33H,4-5,15,17,19H2,1-3H3,(H2,31,35,40)
SMILES CCOC(=O)CCN(C(=O)c1ccc2c(c1)nc(CNc3ccc(cc3)C(=N)NC(=O)OCC)n2C)c4ccccn4
Molecular Weight (g/mol) 571.63
Formula Weight 571.2543
Chemical Name or Material N-Ethoxycarbonyl Dabigatran Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.