missing translation for 'onlineSavingsMsg'
Learn More

9-cis-5,6-Epoxy-5,6-dihydro-retinoic Acid, TRC

Product Code. 30374029
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30374029 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30374029 Supplier Toronto Research Chemicals Supplier No. TRCE5898105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9-cis-5,6-Epoxy-5,6-dihydroretinoic Acid (5,6-Epoxialitretinoine)
CAS 1431303-55-7
Molecular Formula C20 H28 O3
Quantity 5 mg
Synonym 9-cis-5,6-Epoxy-5,6-dihydro-retinoic Acid, Retinoic acid, 5,6-epoxy-5,6-dihydro-, 9-cis-, BAL19222, GSK3040320A, BAL0019222-000, BAL0004079-000, BAL4079, 9-cis retinoic acid, alitretinoin related
InChI Formula InChI=1S/C20H28O3/c1-15(8-6-9-16(2)14-17(21)22)10-13-20-18(3,4)11-7-12-19(20,5)23-20/h6,8-10,13-14H,7,11-12H2,1-5H3,(H,21,22)/b9-6+,13-10+,15-8-,16-14+
SMILES C\C(=C\C=C\C(=C\C(=O)O)\C)\C=C\C12OC1(C)CCCC2(C)C
IUPAC Name (2E,4E,6Z,8E)-3,7-dimethyl-9-(1,5,5-trimethyl-7-oxabicyclo[4.1.0]heptan-6-yl)nona-2,4,6,8-tetraenoic acid
Molecular Weight (g/mol) 316.435
Formula Weight 316.204
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.