missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Maleate, TRC

Product Code. 30374716
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30374716 10 g -
30380795 50 g 50g
30375904 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30374716 Supplier Toronto Research Chemicals Supplier No. TRCD42953510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-76-0
Molecular Formula C12 H20 O4
Molecular Weight (g/mol) 228.28
Synonym 2-Butenedioic acid (2Z)-, dibutyl ester (9CI), 2-Butenedioic acid (Z)-, dibutyl ester, Maleic acid, dibutyl ester (6CI,8CI), Butyl maleate, DBM, Di-n-butyl maleate, Dibutyl maleate, Maleic acid di-n-butyl ester, RC Comonomer DBM, Staflex DBM, Vinavil CF 5
IUPAC Name dibutyl (Z)-but-2-enedioate
SMILES CCCCOC(=O)\C=C/C(=O)OCCCC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H20O4/c1-3-5-9-15-11(13)7-8-12(14)16-10-6-4-2/h7-8H,3-6,9-10H2,1-2H3/b8-7-
Formula Weight 228.1362
Chemical Name or Material Dibutyl maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.