missing translation for 'onlineSavingsMsg'
Learn More

1,9-Diacetoxynonane, TRC

Product Code. 30375768
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30375768 500 mg -
30384604 2.5 g 2.5g
30369105 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30375768 Supplier Toronto Research Chemicals Supplier No. TRCD422443500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4944-60-9
Molecular Formula C13H24O4
Molecular Weight (g/mol) 244.33
IUPAC Name 9-acetyloxynonyl acetate
SMILES CC(=O)OCCCCCCCCCOC(=O)C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C13H24O4/c1-12(14)16-10-8-6-4-3-5-7-9-11-17-13(2)15/h3-11H2,1-2H3
Formula Weight 244.1675
Chemical Name or Material 1,9-Diacetoxynonane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.