missing translation for 'onlineSavingsMsg'
Learn More

1-(Dichloroacetyl)-1,2,3,4-tetrahydro-6-quinolinol, TRC

Product Code. 30376028
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376028 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376028 Supplier Toronto Research Chemicals Supplier No. TRCD43178550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(Dichloroacetyl)-1,2,3,4-tetrahydro-6-quinolinol
CAS 62265-67-2
Purity Grade Notes HPLC
Molecular Formula C11 H11 Cl2 N O2
Quantity 50 mg
Synonym 2,2-Dichloro-1-(3,4-dihydro-6-hydroxy-1(2H)-quinolinyl)ethanone
InChI Formula InChI=1S/C11H11Cl2NO2/c12-10(13)11(16)14-5-1-2-7-6-8(15)3-4-9(7)14/h3-4,6,10,15H,1-2,5H2
SMILES Oc1ccc2N(CCCc2c1)C(=O)C(Cl)Cl
IUPAC Name 2,2-dichloro-1-(6-hydroxy-3,4-dihydro-2H-quinolin-1-yl)ethanone
Molecular Weight (g/mol) 260.12
Formula Weight 259.0167
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.