missing translation for 'onlineSavingsMsg'
Learn More

Dimethylaminomethylfurylmethylthioethyl Ranitidine, TRC

Product Code. 30376112
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376112 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376112 Supplier Toronto Research Chemicals Supplier No. TRCD46098010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N'-Bis[2-[((5-[(dimethylamino)methyl]furan-2-yl)-methyl)sulphanyl]ethyl]-2-nitroethene-1,1-diamine
CAS 72126-78-4
Molecular Formula C22 H35 N5 O4 S2
Quantity 10 mg
Synonym Ranitidine Hydrochloride Imp. A (EP), Ranitidine Imp. A (EP), Ranitidine USP Related Compound B, Ranitidine USP RC B, N,N'-Bis[2-[[[5-[(dimethylamino)methyl]furan-2-yl]-methyl]sulphanyl]ethyl]-2-nitroethene-1,1-diamine, AH 20720X, Ph Eur Ranitidine Impurity A, BP Ranitidine Impurity A, USP Ranitidine Related Compound B, Ranitidine bis-compound (AH20720), Ranitidine Related Compound B, Ranitidine Hydrochloride Impurity A, Ranitidine Impurity A
InChI Formula InChI=1S/C22H35N5O4S2/c1-25(2)13-18-5-7-20(30-18)16-32-11-9-23-22(15-27(28)29)24-10-12-33-17-21-8-6-19(31-21)14-26(3)4/h5-8,15,23-24H,9-14,16-17H2,1-4H3
SMILES CN(C)Cc1oc(CSCCNC(=C[N+](=O)[O-])NCCSCc2oc(CN(C)C)cc2)cc1
IUPAC Name 1-N,1-N'-bis[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethyl]-2-nitroethene-1,1-diamine
Molecular Weight (g/mol) 497.67
Formula Weight 497.213
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.