missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S)-erythro-Dihydro Bupropion Hydrochloride, TRC

Product Code. 30376160
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376160 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376160 Supplier Toronto Research Chemicals Supplier No. TRCD44830525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1R,2S)-erythro-Dihydro Bupropion Hydrochloride
CAS 357628-62-7
Molecular Formula C13 H20 Cl N O . H Cl
Quantity 25 mg
InChI Formula InChI=1S/C13H20ClNO.ClH/c1-9(15-13(2,3)4)12(16)10-6-5-7-11(14)8-10;/h5-9,12,15-16H,1-4H3;1H/t9-,12-;/m0./s1
SMILES Cl.C[C@H](NC(C)(C)C)[C@H](O)c1cccc(Cl)c1
IUPAC Name (1R,2S)-2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-ol;hydrochloride
Molecular Weight (g/mol) 278.218
Formula Weight 277.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.