missing translation for 'onlineSavingsMsg'
Learn More

2-(2,4-Difluorophenyl)-1,2,3-propanetriol, TRC

Product Code. 30376345
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30376345 500 mg
30387867 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30376345 Supplier Toronto Research Chemicals Supplier No. TRCD448105500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 173837-65-5
Molecular Formula C9 H10 F2 O3
Molecular Weight (g/mol) 204.17
Synonym 2-(2,4-Difluorophenyl)propane-1,2,3-triol
IUPAC Name 2-(2,4-difluorophenyl)propane-1,2,3-triol
SMILES OCC(O)(CO)c1ccc(F)cc1F

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H10F2O3/c10-6-1-2-7(8(11)3-6)9(14,4-12)5-13/h1-3,12-14H,4-5H2
Formula Weight 204.0598
Chemical Name or Material 2-(2,4-Difluorophenyl)propane-1,2,3-triol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.