missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-7-(3’carboxy)propoxy-2(1H)-quinolinone, TRC

Product Code. 30376637
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30376637 500 mg
30382455 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30376637 Supplier Toronto Research Chemicals Supplier No. TRCD448725500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58899-27-7
Molecular Formula C13 H15 N O4
Molecular Weight (g/mol) 249.26
Synonym Butanoic acid, 4-[(1,2,3,4-tetrahydro-2-oxo-7-quinolinyl)oxy]-, 4-[(1,2,3,4-Tetrahydro-2-oxo-7-quinolinyl)oxy]butanoic acid, 4-[(2-Oxo-3,4-dihydroquinolin-7(1H)-yl)oxy]butanoic Acid
IUPAC Name 4-[(2-oxo-3,4-dihydro-1H-quinolin-7-yl)oxy]butanoic acid
SMILES OC(=O)CCCOc1ccc2CCC(=O)Nc2c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C13H15NO4/c15-12-6-4-9-3-5-10(8-11(9)14-12)18-7-1-2-13(16)17/h3,5,8H,1-2,4,6-7H2,(H,14,15)(H,16,17)
Formula Weight 249.1001
Chemical Name or Material 3,4-Dihydro-7-(3'carboxy)propoxy-2(1H)-quinolinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.