missing translation for 'onlineSavingsMsg'
Learn More

Ethanesulfonic Acid, TRC

Product Code. 30376686
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30376686 1 g
30376687 100 mg
30369359 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30376686 Supplier Toronto Research Chemicals Supplier No. TRCE9378801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 594-45-6
Molecular Formula C2H6O3S
Molecular Weight (g/mol) 110.13
Synonym 1-Ethanesulfonic acid, Ethylsulfonic acid
SMILES CCS(=O)(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C2H6O3S/c1-2-6(3,4)5/h2H2,1H3,(H,3,4,5)
Formula Weight 110.0038
Chemical Name or Material Ethanesulfonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.