missing translation for 'onlineSavingsMsg'
Learn More

a-Ethyl-benzeneacetyl Chloride, TRC

Product Code. 30377816
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30377816 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30377816 Supplier Toronto Research Chemicals Supplier No. TRCE89975525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha-Ethyl-benzeneacetyl Chloride
CAS 36854-57-6
Molecular Formula C10H11ClO
Quantity 25 g
Synonym (±)-2-Phenylbutyric Chloride, (±)-alpha-Phenylbutyryl Chloride, 2-Phenylbutanoyl Chloride, 2-Phenylbutyryl Chloride, NSC 86133, alpha-Phenylbutyryl Chloride
InChI Formula InChI=1S/C10H11ClO/c1-2-9(10(11)12)8-6-4-3-5-7-8/h3-7,9H,2H2,1H3
SMILES CCC(C(=O)Cl)c1ccccc1
IUPAC Name 2-phenylbutanoyl chloride
Molecular Weight (g/mol) 182.65
Formula Weight 182.0498
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.