missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyldecane, TRC

Product Code. 30378400
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30378400 250 mg -
30387944 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 30378400 Supplier Toronto Research Chemicals Supplier No. TRCD472495250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17302-37-3
Molecular Formula C12H26
Molecular Weight (g/mol) 170.33
Synonym Decane, 2,2-dimethyl-, 2,2-Dimethyldecane
IUPAC Name 2,2-dimethyldecane
SMILES CC(C)(C)CCCCCCCC

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H26/c1-5-6-7-8-9-10-11-12(2,3)4/h5-11H2,1-4H3
Formula Weight 170.2035
Chemical Name or Material 2,2-Dimethyldecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.