missing translation for 'onlineSavingsMsg'
Learn More

5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanone, TRC

Product Code. 30379191
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30379191 5 g
30381119 1 g
30369615 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30379191 Supplier Toronto Research Chemicals Supplier No. TRCE9190055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 698-10-2
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.15
Synonym 2,4-Dihydroxy-3-methyl-2-hexenoic Acid γ-Lactone, 3-Hydroxy-4-methyl-5-ethyl-2(5H)-furanone, Abhexone, Homosotolone, α-Hydroxy-β-methyl-δα,β-γ-hexenolactone, α-Hydroxy-β-methyl-γ-hexenolactone
SMILES O=C1C(O)=C(C)C(CC)O1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H10O3/c1-3-5-4(2)6(8)7(9)10-5/h5,8H,3H2,1-2H3
Formula Weight 142.063
Chemical Name or Material 5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.