missing translation for 'onlineSavingsMsg'
Learn More

(3Z)-2,3-Dihydro-3-[[[4-(methylamino)phenyl]amino]phenylmethylene]-2-oxo-1H-Indole-6-carboxylic Acid Methyl Ester, TRC

Product Code. 30379571
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30379571 2.5 mg 2.5mg
30386017 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30379571 Supplier Toronto Research Chemicals Supplier No. TRCD4924432.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1987887-92-2
Molecular Formula C24 H21 N3 O3
Molecular Weight (g/mol) 399.44
Synonym 1H-Indole-6-carboxylic acid, 2,3-dihydro-3-[[[4-(methylamino)phenyl]amino]phenylmethylene]-2-oxo-, methyl ester, (3Z)- (ACI), Methyl (3Z)-2,3-dihydro-3-[[[4-(methylamino)phenyl]amino]phenylmethylene]-2-oxo-1H-indole-6-carboxylate (ACI), (3Z)-2,3-Dihydro-3-[([4-(methylamino)phenyl]amino)phenylmethylene]-2-oxo-1H-Indole-6-carboxylic Acid Methyl Ester
IUPAC Name methyl (3Z)-3-[[4-(methylamino)anilino]-phenylmethylidene]-2-oxo-1H-indole-6-carboxylate
SMILES CNc1ccc(N\C(=C\2/C(=O)Nc3cc(ccc23)C(=O)OC)\c4ccccc4)cc1

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C24H21N3O3/c1-25-17-9-11-18(12-10-17)26-22(15-6-4-3-5-7-15)21-19-13-8-16(24(29)30-2)14-20(19)27-23(21)28/h3-14,25-26H,1-2H3,(H,27,28)/b22-21-
Formula Weight 399.1583
Chemical Name or Material Nintedanib Impurity D
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.