missing translation for 'onlineSavingsMsg'
Learn More

2,2-dimethoxybutane, TRC

Product Code. 30379577
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30379577 100 mg 100mg
30372627 500 mg -
30385693 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30379577 Supplier Toronto Research Chemicals Supplier No. TRCD473143100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3453-99-4
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.17
IUPAC Name 2,2-dimethoxybutane
SMILES CCC(C)(OC)OC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C6H14O2/c1-5-6(2,7-3)8-4/h5H2,1-4H3
Formula Weight 118.0994
Chemical Name or Material 2,2-dimethoxybutane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.