missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloropyrazolo[3,4-d]pyrimidine, TRC

Product Code. 30380995
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380995 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380995 Supplier Toronto Research Chemicals Supplier No. TRCD4358651G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,6-Dichloropyrazolo[3,4-d]pyrimidine
CAS 42754-96-1
Molecular Formula C5 H2 Cl2 N4
Quantity 1 g
Synonym 4,6-Dichloro-1H-pyrazolo[3,4-d]pyrimidine
InChI Formula InChI=1S/C5H2Cl2N4/c6-3-2-1-8-11-4(2)10-5(7)9-3/h1H,(H,8,9,10,11)
SMILES Clc1nc(Cl)c2cn[nH]c2n1
IUPAC Name 4,6-dichloro-1H-pyrazolo[3,4-d]pyrimidine
Molecular Weight (g/mol) 189
Formula Weight 187.9657
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.