missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dichloropropanoic Acid (>90%), TRC

Product Code. 30381850
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381850 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381850 Supplier Toronto Research Chemicals Supplier No. TRCD4368101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dalapon
CAS 75-99-0
Molecular Formula C3 H4 Cl2 O2
Quantity 1 g
Synonym Propionic acid, 2,2-dichloro- (8CI), 2,2-Dichloropropanoic acid, 2,2-DPA, 2,2-Dichloropropionic acid, Alatex, Basinex P, DPA, Dalapon, Dalascam, NSC 56352, Proprop, S 1315, S 95, S 95 (herbicide), Tripon, α,α-Dichloropropanoic acid, α,α-Dichloropropionic acid
InChI Formula InChI=1S/C3H4Cl2O2/c1-3(4,5)2(6)7/h1H3,(H,6,7)
SMILES CC(Cl)(Cl)C(=O)O
IUPAC Name 2,2-dichloropropanoic acid
Molecular Weight (g/mol) 142.97
Formula Weight 141.9588
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.