missing translation for 'onlineSavingsMsg'
Learn More

Everolimus Retroaldol Degradation Product, TRC

Product Code. 30381891
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381891 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381891 Supplier Toronto Research Chemicals Supplier No. TRCE94542010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Everolimus Retroaldol Degradation Product
CAS 1708118-13-1
Molecular Formula C53H83NO14
Quantity 10 mg
Synonym (S)-(2R,3S,6R,E)-1-((1S,3R,4R)-4-(2-Hydroxyethoxy)-3-methoxycyclohexyl)-2,6,8-trimethyl-5,9-dioxonon-7-en-3-yl 1-(2-((2R,3R,6S)-6-((2S,3E,5E,7E,9S,11R)-2,13-dimethoxy-3,9,11-trimethyl-12-oxotrideca-3,5,7-trien-1-yl)-2-hydroxy-3-methyltetrahydro-2H-pyran-2-yl)-2-oxoacetyl)piperidine-2-carboxylate, Everolimus Ring-opening Impurity
InChI Formula InChI=1S/C53H83NO14/c1-34(26-38(5)45(58)33-63-8)16-12-11-13-17-36(3)47(64-9)30-42-21-19-40(7)53(62,68-42)50(59)51(60)54-23-15-14-18-43(54)52(61)67-48(31-44(57)37(4)27-35(2)32-56)39(6)28-41-20-22-46(66-25-24-55)49(29-41)65-10/h11-13,16-17,27,32,34,37-43,46-49,55,62H,14-15,18-26,28-31,33H2,1-10H3/b13-11+,16-12+,35-27+,36-17+/t34-,37-,38-,39-,40-,41+,42+,43+,46-,47+,48+,49-,53-/m1/s1
SMILES C[C@H](C[C@@]1([H])C[C@@H](OC)[C@H](OCCO)CC1)[C@]([H])(CC([C@H](C)/C=C(C=O)\C)=O)OC([C@H]2N(C(C([C@@]3(O)[C@H](C)CC[C@@]([H])(C[C@H](OC)/C(C)=C/C=C/C=C/[C@@H](C)C[C@@H](C)C(COC)=O)O3)=O)=O)CCCC2)=O
IUPAC Name [(E,2R,3S,6R)-1-[(1S,3R,4R)-4-(2-hydroxyethoxy)-3-methoxycyclohexyl]-2,6,8-trimethyl-5,9-dioxonon-7-en-3-yl] (2S)-1-[2-[(2R,3R,6S)-6-[(2S,3E,5E,7E,9S,11R)-2,13-dimethoxy-3,9,11-trimethyl-12-oxotrideca-3,5,7-trienyl]-2-hydroxy-3-methyloxan-2-yl]-2-oxoacetyl]piperidine-2-carboxylate
Molecular Weight (g/mol) 958.22
Formula Weight 957.5814
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.