missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisobutyl Phthalate-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, bis-iso-butyl ester D4 |
| CAS | 358730-88-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C16 2H4 H18 O4 |
| Quantity | 25 mg |
| Synonym | 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, bis(2-methylpropyl) ester (9CI), Phthalic acid, bis-iso-butyl ester D4, Diisobutyl Phthalate-d4, Phthalic Acid Diisobutyl Ester (3,4,5,6)-D4, Phthalic Acid, Diisobutyl Ester D4 (phenyl D4) |
| InChI Formula | InChI=1S/C16H22O4/c1-11(2)9-19-15(17)13-7-5-6-8-14(13)16(18)20-10-12(3)4/h5-8,11-12H,9-10H2,1-4H3/i5D,6D,7D,8D |
| SMILES | [2H]c1c([2H])c([2H])c(C(=O)OCC(C)C)c(C(=O)OCC(C)C)c1[2H] |
| IUPAC Name | bis(2-methylpropyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 282.37 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?