missing translation for 'onlineSavingsMsg'
Learn More

Diclofenac Levomenthol Ester, TRC

Product Code. 30385420
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385420 25 mg -
30387445 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30385420 Supplier Toronto Research Chemicals Supplier No. TRCD43683025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2648862-09-1
Molecular Formula C24 H29 Cl2 N O2
Molecular Weight (g/mol) 434.399
Synonym (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl 2-(2-((2,6-Dichlorophenyl)amino)phenyl)acetate
IUPAC Name [(1R,2R,5R)-5-methyl-2-propan-2-ylcyclohexyl] 2-[2-(2,6-dichloroanilino)phenyl]acetate
SMILES CC(C)[C@H]1CC[C@@H](C)C[C@H]1OC(=O)Cc2ccccc2Nc3c(Cl)cccc3Cl

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C24H29Cl2NO2/c1-15(2)18-12-11-16(3)13-22(18)29-23(28)14-17-7-4-5-10-21(17)27-24-19(25)8-6-9-20(24)26/h4-10,15-16,18,22,27H,11-14H2,1-3H3/t16-,18-,22-/m1/s1
Formula Weight 433.158
Chemical Name or Material Diclofenac Levomenthol Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.