missing translation for 'onlineSavingsMsg'
Learn More

Diethylammonium Dihydrogen Phosphate, TRC

Product Code. 30388193
Change view
Click to view available options
Quantity:
500 mg
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388193 25 g
30373289 5 g
30376820 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388193 Supplier Toronto Research Chemicals Supplier No. TRCD44375525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68109-72-8
Molecular Formula C4 H11 N . H3 O4 P
Molecular Weight (g/mol) 171.13
Synonym N-Ethylethanamine Phosphate
IUPAC Name N-ethylethanamine;phosphoric acid
SMILES CCNCC.OP(=O)(O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C4H11N.H3O4P/c1-3-5-4-2;1-5(2,3)4/h5H,3-4H2,1-2H3;(H3,1,2,3,4)
Formula Weight 171.066
Percent Purity >95%
Chemical Name or Material Diethylammonium Dihydrogen Phosphate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.