missing translation for 'onlineSavingsMsg'
Learn More

Isobutyryl L-Carnitine Chloride, TRC

Product Code. 30389183
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389183 100 mg
30398854 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30389183 Supplier Toronto Research Chemicals Supplier No. TRCI780500100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6920-31-6
Molecular Formula C11 H22 N O4 . Cl
Molecular Weight (g/mol) 267.75
Synonym 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-(2-methyl-1-oxopropoxy)-, chloride (1:1), (2R)- (ACI), 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-(2-methyl-1-oxopropoxy)-, chloride, (R)- (9CI), Ammonium, (3-carboxy-2-hydroxypropyl)trimethyl-, chloride, isobutyrate, (+)- (8CI), Isobutyryl-L-carnitine Chloride, ST 284, Isobutyryl L-Carnitine Chloride, [(2R)-3-Carboxy-2-(2-methylpropanoyloxy)propyl]trimethylammonium chloride
IUPAC Name [(2R)-3-carboxy-2-(2-methylpropanoyloxy)propyl]-trimethylazanium;chloride
SMILES [Cl-].CC(C)C(=O)O[C@H](CC(=O)O)C[N+](C)(C)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H21NO4.ClH/c1-8(2)11(15)16-9(6-10(13)14)7-12(3,4)5;/h8-9H,6-7H2,1-5H3;1H/t9-;/m1./s1
Formula Weight 267.1237
Chemical Name or Material Isobutyryl L-Carnitine Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.