missing translation for 'onlineSavingsMsg'
Learn More

N5-(5-Hydro-5-methyl-4-imidazolon-2-yl) L-Ornithine Dihydrochloride, TRC

Product Code. 30389604
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389604 5 mg -
30408297 1 mg 1mg
30398415 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389604 Supplier Toronto Research Chemicals Supplier No. TRCH9476305MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C9H16N4O3 . 2HCl
Molecular Weight (g/mol) 301.17
Synonym N5-(4,5-Dihydro-4-methyl-5-oxo-1H-imidazol-2-yl)-L-ornithine Dihydrochloride
IUPAC Name (2S)-2-amino-5-((4-methyl-5-oxo-4,5-dihydro-1H-imidazol-2-yl)amino)pentanoic acid dihydrochloride
SMILES N[C@H](C(O)=O)CCCNC1=NC(C(N1)=O)C.Cl.Cl

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C9H16N4O3.2ClH/c1-5-7(14)13-9(12-5)11-4-2-3-6(10)8(15)16;;/h5-6H,2-4,10H2,1H3,(H,15,16)(H2,11,12,13,14);2*1H/t5?,6-;;/m0../s1
Formula Weight 301.04
Chemical Name or Material N5-(5-Hydro-5-methyl-4-imidazolon-2-yl) L-Ornithine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.