missing translation for 'onlineSavingsMsg'
Learn More

(R)-Alpha-Hydroxycyclohexanebutanoic Acid Ethyl Ester, TRC

Product Code. 30390613
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390613 5 g
30407620 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30390613 Supplier Toronto Research Chemicals Supplier No. TRCH8253705G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95500-39-3
Molecular Formula C12H22O3
Molecular Weight (g/mol) 214.3
Synonym (R)-α-Hydroxycyclohexanebutanoic Acid Ethyl Ester
IUPAC Name ethyl (2R)-4-cyclohexyl-2-hydroxybutanoate
SMILES CCOC(=O)[C@H](O)CCC1CCCCC1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H22O3/c1-2-15-12(14)11(13)9-8-10-6-4-3-5-7-10/h10-11,13H,2-9H2,1H3/t11-/m1/s1
Formula Weight 214.1569
Chemical Name or Material (R)-alpha-Hydroxycyclohexanebutanoic Acid Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.