missing translation for 'onlineSavingsMsg'
Learn More

5-(3'-Hydroxybenzyl)hydantoin, TRC

Product Code. 30391959
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391959 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391959 Supplier Toronto Research Chemicals Supplier No. TRCH8296102G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-(3'-Hydroxybenzyl)hydantoin
CAS 216956-20-6
Molecular Formula C10 H10 N2 O3
Quantity 2 g
InChI Formula InChI=1S/C10H10N2O3/c13-7-3-1-2-6(4-7)5-8-9(14)12-10(15)11-8/h1-4,8,13H,5H2,(H2,11,12,14,15)
SMILES Oc1cccc(CC2NC(=O)NC2=O)c1
IUPAC Name 5-[(3-hydroxyphenyl)methyl]imidazolidine-2,4-dione
Molecular Weight (g/mol) 206.2
Formula Weight 206.0691
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.