missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6beta-Hydroxy Mometasone Furoate, TRC
Specifications
Specifications
| Chemical Name or Material | 6beta-Hydroxy Mometasone Furoate |
| CAS | 132160-74-8 |
| Molecular Formula | C27 H30 Cl2 O7 |
| Quantity | 25 mg |
| Synonym | Pregna-1,4-diene-3,20-dione, 9,21-dichloro-17-[(2-furanylcarbonyl)oxy]-6,11-dihydroxy-16-methyl-, (6β,11β,16α)-, [(6R,8S,9R,10S,11S,13S,14S,16R,17R)-9-chloro-17-(2-chloroacetyl)-6,11-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] furan-2-carboxylate, 6-β-Hydroxy Mometasone Furoate |
| InChI Formula | InChI=1S/C27H30Cl2O7/c1-14-9-16-17-11-19(31)18-10-15(30)6-7-24(18,2)26(17,29)21(32)12-25(16,3)27(14,22(33)13-28)36-23(34)20-5-4-8-35-20/h4-8,10,14,16-17,19,21,31-32H,9,11-13H2,1-3H3/t14-,16+,17+,19-,21+,24+,25+,26+,27+/m1/s1 |
| SMILES | C[C@@H]1C[C@H]2[C@@H]3C[C@@H](O)C4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(OC(=O)c5occc5)C(=O)CCl |
| IUPAC Name | [(6R,8S,9R,10S,11S,13S,14S,16R,17R)-9-chloro-17-(2-chloroacetyl)-6,11-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] furan-2-carboxylate |
| Molecular Weight (g/mol) | 537.43 |
| Formula Weight | 536.1369 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?