missing translation for 'onlineSavingsMsg'
Learn More

JHSB3 , approximately 80%, TRC

Product Code. 30394160
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394160 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394160 Supplier Toronto Research Chemicals Supplier No. TRCJ21103010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material JHSB3
CAS 1198297-86-7
Molecular Formula C16H26O4
Quantity 10 mg
Synonym (2R,3S)-3-[(3E)-6-[(2R)-3,3-Dimethyl-2-oxiranyl]-4-methyl-3-hexen-1-yl]-3-methyl-2-oxiranecarboxylic Acid Methyl Ester, Juvenile Hormone SB3
InChI Formula InChI=1S/C16H26O4/c1-11(8-9-12-15(2,3)19-12)7-6-10-16(4)13(20-16)14(17)18-5/h7,12-13H,6,8-10H2,1-5H3/b11-7+/t12-,13+,16+/m1/s1
SMILES Cc1cc2c3c(cccc3)nc2nc1
Molecular Weight (g/mol) 282.38
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.