missing translation for 'onlineSavingsMsg'
Learn More

Fidaxomicin, TRC

Product Code. 30394437
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394437 5 mg
30395055 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394437 Supplier Toronto Research Chemicals Supplier No. TRCF3354005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873857-62-6
Molecular Formula C52 H74 Cl2 O18
Molecular Weight (g/mol) 1058.04
Synonym Oxacyclooctadeca-3,5,9,13,15-pentaen-2-one, 3-[[[6-deoxy-4-O-(3,5-dichloro-2-ethyl-4,6-dihydroxybenzoyl)-2-O-methyl-β-D-mannopyranosyl]oxy]methyl]-12-[[6-deoxy-5-C-methyl-4-O-(2-methyl-1-oxopropyl)-β-D-lyxo-hexopyranosyl]oxy]-11-ethyl-8-hydroxy-18-[(1R)-1-hydroxyethyl]-9,13,15-trimethyl-, (3E,5E,8S,9E,11S,12R,13E,15E,18S)-, (3E,5E,8S,9E,11S,12R,13E,15E,18S)-3-[[[6-Deoxy-4-O-(3,5-dichloro-2-ethyl-4,6-dihydroxybenzoyl)-2-O-methyl-β-D-mannopyranosyl]oxy]methyl]-12-[[6-deoxy-5-C-methyl-4-O-(2-methyl-1-oxopropyl)-β-D-lyxo-hexopyranosyl]oxy]-11-ethyl-8-hydroxy-18-[(1R)-1-hydroxyethyl]-9,13,15-trimethyloxacyclooctadeca-3,5,9,13,15-pentaen-2-one, Clostomicin B1, Dificid, Dificlir, Fidaxomicin, Fidaxomycin, Lipiarmicin, Lipiarmycin, Lipiarmycin A 3, OPT 80, PAR 01, PAR 101, R-Tiacumicin B, Tiacumicin B
IUPAC Name [(2R,3S,4S,5S,6R)-6-[[(3E,5E,8S,9E,11S,12R,13E,15E,18S)-12-[(2R,3S,4R,5S)-3,4-dihydroxy-6,6-dimethyl-5-(2-methylpropanoyloxy)oxan-2-yl]oxy-11-ethyl-8-hydroxy-18-[(1R)-1-hydroxyethyl]-9,13,15-trimethyl-2-oxo-1-oxacyclooctadeca-3,5,9,13,15-pentaen-3-yl]methoxy]-4-hydroxy-5-methoxy-2-methyloxan-3-yl] 3,5-dichloro-2-ethyl-4,6-dihydroxybenzoate
SMILES CC[C@H]1\C=C(/C)\[C@@H](O)C\C=C\C=C(/CO[C@@H]2O[C@H](C)[C@@H](OC(=O)c3c(O)c(Cl)c(O)c(Cl)c3CC)[C@H](O)[C@@H]2OC)\C(=O)O[C@@H](C\C=C(/C)\C=C(/C)\[C@@H]1O[C@@H]4OC(C)(C)[C@@H](OC(=O)C(C)C)[C@H](O)[C@@H]4O)[C@@H](C)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C52H74Cl2O18/c1-13-30-22-26(6)33(56)18-16-15-17-31(23-66-51-45(65-12)42(61)44(29(9)67-51)69-49(64)35-32(14-2)36(53)39(58)37(54)38(35)57)48(63)68-34(28(8)55)20-19-25(5)21-27(7)43(30)70-50-41(60)40(59)46(52(10,11)72-50)71-47(62)24(3)4/h15-17,19,21-22,24,28-30,33-34,40-46,50-51,55-61H,13-14,18,20,23H2,1-12H3/b16-15+,25-19+,26-22+,27-21+,31-17+/t28-,29-,30+,33+,34+,40-,41+,42+,43+,44-,45+,46+,50-,51-/m1/s1
Formula Weight 1056.4252
Percent Purity >95%
Chemical Name or Material Fidaxomicin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.